Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013820.3 | NP_038848.1 | 17 - 18 | 3082 | 61 |
GenBank mRNA | AJ238540.1 | - | 17 - 18 | 3025 | 61 |
AK145936.1 | - | 17 - 18 | 3062 | 61 | |
BC054472.1 | - | 17 - 18 | 3001 | 61 |