Assay Design



Gene Transcripts

Gene Symbol
Hk2
Entrez Gene ID
15277
Gene Name
hexokinase 2
Gene Aliases
AI642394, HKII
Chromosome Location
Chr.6: 82725025 - 82774454 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013820.3 NP_038848.1 1 - 2 533 64
GenBank mRNA AJ238540.1 - 1 - 2 476 64
AK145936.1 - 1 - 2 514 64
BC054472.1 - 1 - 2 452 64

View Full Product Details