Assay Design



Gene Transcripts

Gene Symbol
Hk2
Entrez Gene ID
15277
Gene Name
hexokinase 2
Gene Aliases
AI642394, HKII
Chromosome Location
Chr.6: 82725025 - 82774454 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013820.3 NP_038848.1 5 - 6 1065 59
GenBank mRNA AJ238540.1 - 5 - 6 1008 59
AK145936.1 - 5 - 6 1046 59
BC054472.1 - 5 - 6 984 59

View Full Product Details