Assay Design



Gene Transcripts

Gene Symbol
Krcc1
Entrez Gene ID
57896
Gene Name
lysine-rich coiled-coil 1
Gene Aliases
AA792894
Chromosome Location
Chr.6: 71271569 - 71285319 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK050640.1 - 2 - 3 321 145
AK146818.1 - 2 - 3 332 145
AK148096.1 - 2 - 3 311 145
AK162678.1 - 2 - 3 321 145
BC046798.1 - 2 - 3 289 145

View Full Product Details