Assay Design



Gene Transcripts

Gene Symbol
Kbtbd8
Entrez Gene ID
243574
Gene Name
kelch repeat and BTB (POZ) domain containing 8
Gene Aliases
AU040152, SSEC-51, SSEC51, Takrp, mKIAA1842
Chromosome Location
Chr.6: 95117880 - 95129793 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001008785.5 NP_001008785.2 1 - 2 95 83
GenBank mRNA AK142080.1 - 1 - 2 69 83

View Full Product Details