Assay Design



Gene Transcripts

Gene Symbol
Lmod3
Entrez Gene ID
320502
Gene Name
leiomodin 3 (fetal)
Gene Aliases
5430424A14Rik
Chromosome Location
Chr.6: 97238528 - 97253097 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081157.1 NP_001074626.1 1 - 2 504 77
GenBank mRNA AK030676.1 - 1 - 2 484 77
AK052890.1 - 1 - 2 417 77
BC127157.1 - 1 - 2 435 77

View Full Product Details