Assay Design
Gene Transcripts
- Gene Symbol
- March8
- Entrez Gene ID
-
71779
- Gene Name
- membrane-associated ring finger (C3HC4) 8
- Gene Aliases
-
1300017E09Rik, MARCH-VIII, Mir
- Chromosome Location
-
Chr.6: 116337884 - 116409540 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK005032.2 |
- |
1 - 2 |
103 |
71 |
BC050908.1 |
- |
2 - 3 |
278 |
71 |
BC053090.1 |
- |
1 - 2 |
363 |
71 |
View Full Product Details