Assay Design



Gene Transcripts

Gene Symbol
Il17ra
Entrez Gene ID
16172
Gene Name
interleukin 17 receptor A
Gene Aliases
AW538159, Cdw217, Il17r, VDw217
Chromosome Location
Chr.6: 120463181 - 120483731 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008359.2 NP_032385.1 3 - 4 443 67
GenBank mRNA AK080226.2 - 3 - 4 394 67
U31993.1 - 3 - 4 431 67

View Full Product Details