Assay Design



Gene Transcripts

Gene Symbol
Kdm5a
Entrez Gene ID
214899
Gene Name
lysine (K)-specific demethylase 5A
Gene Aliases
AA409370, C76986, Jarid1a, RBP2, Rbbp2
Chromosome Location
Chr.6: 120364099 - 120444574 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145997.2 NP_666109.2 16 - 17 2792 70
GenBank mRNA AK135085.1 - 1 - 2 120 70
AK144877.1 - 16 - 17 2763 70
AK159079.1 - 16 - 17 2769 70
AK166055.1 - 16 - 17 2773 70
BC010717.1 - 16 - 17 2458 70
BC022581.1 - 1 - 2 67 70
BC080691.1 - 16 - 17 2478 70

View Full Product Details