Assay Design



Gene Transcripts

Gene Symbol
Kdm5a
Entrez Gene ID
214899
Gene Name
lysine (K)-specific demethylase 5A
Gene Aliases
AA409370, C76986, Jarid1a, RBP2, Rbbp2
Chromosome Location
Chr.6: 120364099 - 120444574 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145997.2 NP_666109.2 26 - 27 4973 105
GenBank mRNA AK134200.1 - 8 - 9 1791 105
AK135085.1 - 11 - 12 2301 105

View Full Product Details