Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_145997.2 | NP_666109.2 | 7 - 8 | 1379 | 72 |
GenBank mRNA | AK144877.1 | - | 7 - 8 | 1350 | 72 |
AK159079.1 | - | 7 - 8 | 1356 | 72 | |
AK166055.1 | - | 7 - 8 | 1360 | 72 | |
BC010717.1 | - | 7 - 8 | 1020 | 72 | |
BC011506.3 | - | 7 - 8 | 1313 | 72 | |
BC080691.1 | - | 7 - 8 | 1065 | 72 |