Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009531.2 | NP_033557.2 | 13 - 14 | 2485 | 84 |
GenBank mRNA | AB071144.1 | - | 13 - 14 | 2422 | 84 |
AK004713.1 | - | 13 - 14 | 2425 | 84 | |
AK028595.1 | - | 13 - 14 | 2423 | 84 | |
AK166981.1 | - | 13 - 14 | 2420 | 84 | |
AK170975.1 | - | 12 - 13 | 4278 | 84 | |
BC052725.1 | - | 13 - 14 | 2447 | 84 |