Assay Design



Gene Transcripts

Gene Symbol
Xpc
Entrez Gene ID
22591
Gene Name
xeroderma pigmentosum, complementation group C
Gene Aliases
-
Chromosome Location
Chr.6: 91489305 - 91515888 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009531.2 NP_033557.2 7 - 8 971 67
GenBank mRNA AB071144.1 - 7 - 8 908 67
AK004713.1 - 7 - 8 911 67
AK028595.1 - 7 - 8 909 67
AK166981.1 - 7 - 8 906 67
AK170975.1 - 7 - 8 903 67
BC052725.1 - 7 - 8 933 67
U27398.1 - 7 - 8 879 67
U40005.1 - 6 - 7 710 67

View Full Product Details