Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025823.4 | NP_080099.1 | 2 - 3 | 384 | 97 |
GenBank mRNA | AK004799.2 | - | 2 - 3 | 339 | 97 |
AK004840.1 | - | 2 - 3 | 339 | 97 | |
AK031585.1 | - | 2 - 3 | 340 | 97 | |
AK033373.1 | - | 2 - 3 | 340 | 97 | |
AK049412.1 | - | 2 - 3 | 337 | 97 | |
AK134528.1 | - | 2 - 3 | 385 | 97 | |
AK145366.1 | - | 2 - 3 | 344 | 97 | |
AK147190.1 | - | 2 - 3 | 352 | 97 | |
AK159981.1 | - | 2 - 3 | 340 | 97 | |
AK161616.1 | - | 2 - 3 | 340 | 97 | |
AK169209.1 | - | 2 - 3 | 340 | 97 | |
AK173058.1 | - | 2 - 3 | 306 | 97 | |
BC028308.1 | - | 2 - 3 | 354 | 97 |