Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011818.3 | NP_035948.3 | 13 - 14 | 1657 | 65 |
GenBank mRNA | AF163665.1 | - | 13 - 14 | 1601 | 65 |
AF186095.1 | - | 13 - 14 | 1493 | 65 | |
AF282322.1 | - | 13 - 14 | 1527 | 65 | |
AK004716.1 | - | 13 - 14 | 1633 | 65 | |
AK146964.1 | - | 13 - 14 | 1626 | 65 | |
AK155238.1 | - | 13 - 14 | 1640 | 65 | |
BC054767.1 | - | 13 - 14 | 1616 | 65 |