Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_013471.2 | NP_038499.2 | 9 - 10 | 739 | 73 |
GenBank mRNA | AK132293.1 | - | 10 - 11 | 781 | 73 |
AK150486.1 | - | 9 - 10 | 700 | 73 | |
AK150614.1 | - | 9 - 10 | 686 | 73 | |
AK151054.1 | - | 9 - 10 | 686 | 73 | |
AK151236.1 | - | 9 - 10 | 718 | 73 | |
AK167338.1 | - | 9 - 10 | 740 | 73 | |
AK168390.1 | - | 9 - 10 | 711 | 73 | |
AK168487.1 | - | 9 - 10 | 720 | 73 | |
AK168917.1 | - | 8 - 9 | 744 | 73 | |
AK170447.1 | - | 9 - 10 | 685 | 73 | |
BC055871.1 | - | 9 - 10 | 699 | 73 | |
U72941.1 | - | 9 - 10 | 710 | 73 | |
U95371.1 | - | 10 - 11 | 1398 | 73 |