Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019948.2 | NP_064332.1 | 5 - 6 | 610 | 92 |
GenBank mRNA | AB024717.1 | - | 5 - 6 | 610 | 92 |
AK036493.1 | - | 4 - 5 | 2986 | 92 | |
AK089286.1 | - | 4 - 5 | 681 | 92 | |
AK150528.1 | - | 5 - 6 | 610 | 92 | |
AK150899.1 | - | 5 - 6 | 610 | 92 | |
AK151101.1 | - | 5 - 6 | 609 | 92 | |
AK151912.1 | - | 5 - 6 | 609 | 92 | |
AK152333.1 | - | 3 - 4 | 3059 | 92 | |
AK153008.1 | - | 5 - 6 | 609 | 92 | |
AK153308.1 | - | 5 - 6 | 610 | 92 | |
AK155452.1 | - | 4 - 5 | 682 | 92 | |
AK156437.1 | - | 5 - 6 | 610 | 92 | |
AK170698.1 | - | 5 - 6 | 561 | 92 | |
AK171103.1 | - | 5 - 6 | 610 | 92 | |
BC003218.1 | - | 5 - 6 | 568 | 92 | |
CT010223.1 | - | 5 - 6 | 487 | 92 |