Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033253.3 | NP_001028425.1 | 15 - 16 | 3283 | 99 |
NM_001159942.1 | NP_001153414.1 | 16 - 17 | 3449 | 99 | |
GenBank mRNA | AK122468.1 | - | 12 - 13 | 2607 | 99 |
AK135923.1 | - | 15 - 16 | 3285 | 99 | |
BC076628.1 | - | 14 - 15 | 2645 | 99 | |
BC117965.1 | - | 14 - 15 | 2885 | 99 | |
BC117966.1 | - | 14 - 15 | 2878 | 99 |