Assay Design



Gene Transcripts

Gene Symbol
Plekhg1
Entrez Gene ID
213783
Gene Name
pleckstrin homology domain containing, family G (with RhoGef domain) member 1
Gene Aliases
D10Ertd733e, Gm521, mKIAA1209
Chromosome Location
Chr.10: 3740251 - 3967303 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033253.3 NP_001028425.1 6 - 7 935 75
NM_001159942.1 NP_001153414.1 7 - 8 1101 75
GenBank mRNA AK033676.1 - 7 - 8 1101 75
AK034403.1 - 7 - 8 1102 75
AK045140.1 - 7 - 8 1281 75
AK122468.1 - 3 - 4 259 75
AK135923.1 - 6 - 7 937 75
BC076628.1 - 4 - 5 332 75
BC117965.1 - 5 - 6 537 75
BC117966.1 - 5 - 6 530 75

View Full Product Details