Assay Design



Gene Transcripts

Gene Symbol
Plekhg1
Entrez Gene ID
213783
Gene Name
pleckstrin homology domain containing, family G (with RhoGef domain) member 1
Gene Aliases
D10Ertd733e, Gm521, mKIAA1209
Chromosome Location
Chr.10: 3740251 - 3967303 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001033253.3 NP_001028425.1 10 - 11 1450 83
NM_001159942.1 NP_001153414.1 11 - 12 1616 83
GenBank mRNA AK122468.1 - 7 - 8 774 83
AK135923.1 - 10 - 11 1452 83
BC076628.1 - 8 - 9 847 83
BC117965.1 - 9 - 10 1052 83
BC117966.1 - 9 - 10 1045 83

View Full Product Details