Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_053204.2 | NP_444434.2 | 20 - 21 | 3791 | 64 |
NR_104484.1 | 20 - 21 | 3726 | 64 | ||
GenBank mRNA | AF340028.1 | - | 20 - 21 | 3710 | 64 |
AK122437.1 | - | 9 - 10 | 1089 | 64 |