Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011401.4 | NP_035531.3 | 9 - 10 | 1625 | 79 |
GenBank mRNA | AK028041.1 | - | 9 - 10 | 1401 | 79 |
AK147258.1 | - | 8 - 9 | 3937 | 79 | |
AK155997.1 | - | 9 - 10 | 1384 | 79 | |
AK164286.1 | - | 9 - 10 | 1397 | 79 | |
BC034122.1 | - | 9 - 10 | 1383 | 79 | |
BC050095.1 | - | 9 - 10 | 1379 | 79 | |
BC058811.1 | - | 9 - 10 | 1383 | 79 | |
CT010354.1 | - | 8 - 9 | 1270 | 79 | |
M75135.1 | - | 9 - 10 | 1349 | 79 | |
X61093.1 | - | 9 - 10 | 1349 | 79 |