Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008021.4 | NP_032047.4 | 7 - 8 | 1256 | 75 |
GenBank mRNA | AK049177.1 | - | 7 - 8 | 1197 | 75 |
BC065067.1 | - | 7 - 8 | 1256 | 75 | |
BC075723.1 | - | 7 - 8 | 1275 | 75 | |
Y11245.1 | - | 7 - 8 | 1164 | 75 |