Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033463.3 | NP_001028635.2 | 5 - 6 | 1959 | 80 |
GenBank mRNA | AK146408.1 | - | 4 - 5 | 1891 | 80 |
AK156197.1 | - | 5 - 6 | 1960 | 80 | |
AK220217.1 | - | 4 - 5 | 2073 | 80 | |
BC141065.1 | - | 5 - 6 | 2077 | 80 | |
BC145318.1 | - | 5 - 6 | 2077 | 80 |