Assay Design



Gene Transcripts

Gene Symbol
Ankrd53
Entrez Gene ID
75305
Gene Name
ankyrin repeat domain 53
Gene Aliases
4930564N15Rik
Chromosome Location
Chr.6: 83762646 - 83768326 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029245.3 NP_083521.2 3 - 4 753 105
GenBank mRNA AK016222.1 - 2 - 3 549 105
AK133099.1 - 3 - 4 755 105

View Full Product Details