Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_181470.4 | NP_852135.1 | 10 - 11 | 1409 | 150 |
GenBank mRNA | AK011491.1 | - | 10 - 11 | 1395 | 150 |
BC067062.1 | - | 10 - 11 | 1409 | 150 | |
BC069962.1 | - | 10 - 11 | 1403 | 150 | |
U01139.1 | - | 7 - 8 | 984 | 150 |