Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026257.3 | NP_080533.1 | 1 - 2 | 68 | 56 |
NR_045571.1 | 1 - 2 | 68 | 56 | ||
GenBank mRNA | AK015720.1 | - | 1 - 2 | 52 | 56 |
AK048494.1 | - | 1 - 2 | 69 | 56 | |
BC019461.1 | - | 1 - 2 | 45 | 56 |