Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039677.2 | NP_001034766.1 | 2 - 3 | 501 | 56 |
GenBank mRNA | AK031425.1 | - | 2 - 3 | 297 | 56 |
BC013478.1 | - | 1 - 2 | 521 | 56 | |
BC113751.1 | - | 2 - 3 | 501 | 56 | |
BC147704.1 | - | 2 - 3 | 332 | 56 | |
BC147707.1 | - | 2 - 3 | 332 | 56 |