Assay Design
Gene Transcripts
- Gene Symbol
- Slc30a2
- Entrez Gene ID
-
230810
- Gene Name
- solute carrier family 30 (zinc transporter), member 2
- Gene Aliases
-
6030430C21, ZnT-2, Znt2
- Chromosome Location
-
Chr.4: 134342961 - 134356528 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC013478.1 |
- |
2 - 3 |
670 |
71 |
BC147704.1 |
- |
3 - 4 |
481 |
71 |
BC147707.1 |
- |
3 - 4 |
481 |
71 |
View Full Product Details