Assay Design



Gene Transcripts

Gene Symbol
Tigd2
Entrez Gene ID
68140
Gene Name
tigger transposable element derived 2
Gene Aliases
3632410O17Rik
Chromosome Location
Chr.6: 59208870 - 59212033 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001081145.1 NP_001074614.1 1 - 2 312 111
GenBank mRNA AK005391.1 - 1 - 2 190 111
AK019456.1 - 1 - 2 206 111

View Full Product Details