Assay Design



Gene Transcripts

Gene Symbol
Grm1
Entrez Gene ID
14816
Gene Name
glutamate receptor, metabotropic 1
Gene Aliases
4930455H15Rik, ENSMUSG00000075319, Gm10828, Gprc1a, mGluR1, nmf373, rcw, wobl
Chromosome Location
Chr.10: 10686059 - 11082377 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001114333.2 NP_001107805.1 1 - 2 86 68
NM_016976.3 NP_058672.1 1 - 2 86 68
GenBank mRNA AF320126.2 - 1 - 2 111 68
BC067057.1 - 1 - 2 60 68
CJ129468.1 - 1 - 2 87 68

View Full Product Details