Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001114333.2 | NP_001107805.1 | 6 - 7 | 2003 | 61 |
NM_016976.3 | NP_058672.1 | 6 - 7 | 2003 | 61 | |
GenBank mRNA | AF320126.2 | - | 6 - 7 | 2028 | 61 |
AK132901.1 | - | 2 - 3 | 233 | 61 | |
BC067057.1 | - | 6 - 7 | 1977 | 61 | |
BC079566.1 | - | 5 - 6 | 2020 | 61 |