Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_172481.2 | NP_766069.1 | 2 - 3 | 119 | 103 |
GenBank mRNA | AK054426.1 | - | 2 - 3 | 120 | 103 |
AY355342.1 | - | 2 - 3 | 120 | 103 | |
BC066795.1 | - | 2 - 3 | 129 | 103 |