Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111277.1 | NP_001104747.1 | 2 - 3 | 231 | 63 |
NM_175135.4 | NP_780344.2 | 2 - 3 | 231 | 63 | |
GenBank mRNA | AK145856.1 | - | 2 - 3 | 208 | 63 |
AK172173.1 | - | 2 - 3 | 225 | 63 | |
AK172302.1 | - | 2 - 3 | 181 | 63 |