Assay Design



Gene Transcripts

Gene Symbol
Chmp2a
Entrez Gene ID
68953
Gene Name
charged multivesicular body protein 2A
Gene Aliases
1500016L11Rik
Chromosome Location
Chr.7: 13032006 - 13034777 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026885.3 NP_081161.1 2 - 3 357 102
GenBank mRNA AK005267.1 - 2 - 3 325 102
BC012230.1 - 2 - 3 298 102

View Full Product Details