Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008698.2 | NP_032724.1 | 7 - 8 | 614 | 68 |
GenBank mRNA | AJ001260.1 | - | 7 - 8 | 614 | 68 |
AK013140.1 | - | 7 - 8 | 622 | 68 | |
AK031804.1 | - | 7 - 8 | 630 | 68 | |
AK147786.1 | - | 8 - 9 | 891 | 68 | |
BC010837.1 | - | 7 - 8 | 612 | 68 |