Assay Design



Gene Transcripts

Gene Symbol
Tmem53
Entrez Gene ID
68777
Gene Name
transmembrane protein 53
Gene Aliases
1110038M16Rik, 1500001P22Rik, 1700012A05Rik
Chromosome Location
Chr.4: 117251951 - 117268588 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_026837.3 NP_081113.1 1 - 2 180 80
GenBank mRNA AK004164.1 - 1 - 2 143 80
BC019937.1 - 1 - 2 106 80

View Full Product Details