Assay Design



Gene Transcripts

Gene Symbol
Plagl1
Entrez Gene ID
22634
Gene Name
pleiomorphic adenoma gene-like 1
Gene Aliases
Lot1, Zac1
Chromosome Location
Chr.10: 13090788 - 13131694 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009538.2 NP_033564.2 1 - 2 238 61
GenBank mRNA AK142210.1 - 1 - 2 196 61
BC065150.1 - 1 - 2 238 61

View Full Product Details