Assay Design
Gene Transcripts
- Gene Symbol
- Map7
- Entrez Gene ID
-
17761
- Gene Name
- microtubule-associated protein 7
- Gene Aliases
-
E-MAP-115, MAP-7, Mtap7, R75000, mshi, mste, ste
- Chromosome Location
-
Chr.10: 20148920 - 20281590 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AK008018.1 |
- |
1 - 2 |
107 |
79 |
View Full Product Details