Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001145970.1 | NP_001139442.1 | 11 - 12 | 2085 | 144 |
NM_144941.3 | NP_659190.3 | 11 - 12 | 2301 | 144 | |
GenBank mRNA | AK122462.1 | - | 5 - 6 | 3724 | 144 |
AK151613.1 | - | 9 - 10 | 1816 | 144 | |
BC016081.1 | - | 2 - 3 | 337 | 144 | |
BC019977.1 | - | 6 - 7 | 1277 | 144 | |
BC023677.1 | - | 11 - 12 | 2276 | 144 | |
BC023774.1 | - | 11 - 12 | 2082 | 144 | |
BM944422.1 | - | 2 - 3 | 191 | 144 |