Assay Design



Gene Transcripts

Gene Symbol
Dguok
Entrez Gene ID
27369
Gene Name
deoxyguanosine kinase
Gene Aliases
dGK, dGK1, dGK2, dGK3
Chromosome Location
Chr.6: 83480214 - 83506969 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162521.1 NP_001155993.1 5 - 6 784 89
GenBank mRNA AY037862.1 - 5 - 6 714 89
BC094920.1 - 5 - 6 784 89

View Full Product Details