Assay Design



Gene Transcripts

Gene Symbol
Ccdc77
Entrez Gene ID
67200
Gene Name
coiled-coil domain containing 77
Gene Aliases
2400002C23Rik, 2700091N06Rik, AV168274
Chromosome Location
Chr.6: 120324322 - 120364369 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_045558.1 2 - 3 339 98
GenBank mRNA AK010251.1 - 2 - 3 106 97
AK031590.1 - 2 - 3 340 98
BC018387.1 - 2 - 3 80 98

View Full Product Details