Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_025958.2 | NP_080234.2 | 6 - 7 | 921 | 64 |
GenBank mRNA | AK129186.1 | - | 6 - 7 | 892 | 64 |
AK145112.1 | - | 6 - 7 | 906 | 64 | |
BC056365.1 | - | 6 - 7 | 903 | 64 | |
BC058338.1 | - | 5 - 6 | 736 | 64 |