Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_030165.3 | NP_084441.3 | 6 - 7 | 1630 | 96 |
GenBank mRNA | AK019496.1 | - | 6 - 7 | 1633 | 96 |
AK028045.1 | - | 6 - 7 | 1611 | 96 | |
AK145992.1 | - | 6 - 7 | 1617 | 96 | |
AK160867.1 | - | 6 - 7 | 1623 | 96 | |
BC023112.1 | - | 2 - 3 | 112 | 96 | |
BC060169.1 | - | 4 - 5 | 3233 | 96 |