Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009819.2 | NP_033949.2 | 1 - 2 | 252 | 107 |
GenBank mRNA | AK035033.1 | - | 1 - 2 | 148 | 107 |
BC051995.1 | - | 1 - 2 | 135 | 107 | |
CD578966.1 | - | 1 - 2 | 252 | 107 | |
D25282.1 | - | 1 - 2 | 181 | 107 |