Assay Design



Gene Transcripts

Gene Symbol
Alms1
Entrez Gene ID
236266
Gene Name
Alstrom syndrome 1
Gene Aliases
bbb
Chromosome Location
Chr.6: 85587499 - 85726611 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_145223.2 NP_660258.2 12 - 13 7415 74
GenBank mRNA AF425257.1 - 12 - 13 7416 74

View Full Product Details