Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001034029.1 | NP_001029201.1 | 9 - 10 | 936 | 62 |
NM_001034031.1 | NP_001029203.1 | 11 - 12 | 1011 | 62 | |
NM_145826.5 | NP_665825.2 | 10 - 11 | 1363 | 62 | |
GenBank mRNA | AF458068.1 | - | 10 - 11 | 1013 | 62 |
AK046302.1 | - | 10 - 11 | 1366 | 62 | |
AK046713.1 | - | 10 - 11 | 1025 | 62 | |
AK169498.1 | - | 10 - 11 | 1191 | 62 | |
BC069861.1 | - | 10 - 11 | 1040 | 62 | |
DQ092339.1 | - | 10 - 11 | 952 | 62 | |
DQ092340.1 | - | 9 - 10 | 936 | 62 | |
DQ092341.1 | - | 11 - 12 | 1011 | 62 |