Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175313.4 | NP_780522.1 | 12 - 13 | 1501 | 75 |
GenBank mRNA | AK037507.1 | - | 12 - 13 | 1502 | 75 |
AK205004.1 | - | 2 - 3 | 106 | 75 | |
BC046414.1 | - | 6 - 7 | 564 | 75 | |
BC048939.1 | - | 10 - 11 | 1040 | 75 |