Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001001160.3 | NP_001001160.1 | 12 - 13 | 3046 | 62 |
NM_001289674.1 | NP_001276603.1 | 11 - 12 | 2877 | 62 | |
NM_001289675.1 | NP_001276604.1 | 11 - 12 | 2871 | 62 | |
GenBank mRNA | AK129466.1 | - | 10 - 11 | 2423 | 62 |
AK147230.1 | - | 12 - 13 | 2938 | 62 | |
AK147681.1 | - | 11 - 12 | 2736 | 62 | |
BC061475.1 | - | 9 - 10 | 1518 | 62 | |
BC070445.1 | - | 11 - 12 | 2877 | 62 |