Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177412.1 | NP_803131.1 | 5 - 6 | 1932 | 85 |
GenBank mRNA | AK028322.1 | - | 3 - 4 | 1561 | 85 |
AK154540.1 | - | 3 - 4 | 1618 | 85 | |
AK173022.1 | - | 5 - 6 | 1921 | 85 | |
BC029759.1 | - | 4 - 5 | 1754 | 85 | |
BC157922.1 | - | 4 - 5 | 1855 | 85 |